Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536790.6 | XP_536790.2 | 4 - 5 | 413 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |