Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIH1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619775.3 | XP_005619832.1 | 604 | 71 | ||
XM_022418309.1 | XP_022274017.1 | 810 | 71 | |||
XM_022418310.1 | XP_022274018.1 | 1 | 263 | 71 | ||
XM_022418312.1 | XP_022274020.1 | 604 | 71 | |||
XM_022418313.1 | XP_022274021.1 | 589 | 71 | |||
XM_536575.6 | XP_536575.1 | 584 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |