Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434684.4 | XP_003434732.1 | 356 | 62 | ||
XM_005620570.3 | XP_005620627.1 | 380 | 62 | |||
XM_005620571.3 | XP_005620628.1 | 346 | 62 | |||
XM_014113950.2 | XP_013969425.1 | 379 | 62 | |||
XM_844929.5 | XP_850022.1 | 413 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris |