Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115175.2 | XP_013970650.1 | 2 - 3 | 293 | 80 | |
XM_022421023.1 | XP_022276731.1 | 2 - 3 | 201 | 80 | ||
XM_537182.6 | XP_537182.1 | 2 - 3 | 291 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |