Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423297.1 | XP_022279005.1 | 1 - 2 | 195 | 55 | |
XM_537722.6 | XP_537722.1 | 1 - 2 | 186 | 55 |
Set Membership: |
![]() ![]() ![]() ![]() |