Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536603.6 | XP_536603.2 | 2 - 3 | 1528 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |