Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC480425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623772.3 | XP_005623829.1 | 3 - 4 | 1030 | 72 | |
XM_537546.6 | XP_537546.2 | 3 - 4 | 1070 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |