Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417626.1 | XP_022273334.1 | 9 - 10 | 879 | 89 | |
XM_022417627.1 | XP_022273335.1 | 9 - 10 | 868 | 89 | ||
XM_536412.6 | XP_536412.2 | 8 - 9 | 776 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |