Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536709.6 | XP_536709.1 | 354 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris |