Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118409.2 | XP_013973884.1 | 4 - 5 | 389 | 120 | |
XM_022426011.1 | XP_022281719.1 | 3 - 4 | 593 | 120 | ||
XM_538897.6 | XP_538897.3 | 3 - 4 | 359 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |