Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_541942.6 | XP_541942.2 | 1 - 2 | 171 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |