Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_541624.6 | XP_541624.3 | 1 - 2 | 122 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |