Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627299.2 | XP_005627356.1 | 3 - 4 | 349 | 70 | |
XM_022425769.1 | XP_022281477.1 | 3 - 4 | 433 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |