Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_543123.6 | XP_543123.3 | 932 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris |