Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544652.6 | XP_544652.3 | 3 - 4 | 1875 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |