Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022407989.1 | XP_022263697.1 | 2562 | 89 | ||
XM_544263.6 | XP_544263.2 | 2765 | 89 |