Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544054.6 | XP_544054.2 | 4 - 5 | 665 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |