Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619188.3 | XP_005619245.1 | 1 - 2 | 216 | 94 | |
XM_005619189.3 | XP_005619246.1 | 1 - 2 | 126 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |