Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619187.3 | XP_005619244.1 | 2 - 3 | 212 | 78 | |
XM_005619188.3 | XP_005619245.1 | 2 - 3 | 340 | 78 | ||
XM_005619189.3 | XP_005619246.1 | 2 - 3 | 250 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |