Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417442.1 | XP_022273150.1 | 2 - 3 | 776 | 136 | |
XM_022417443.1 | XP_022273151.1 | 2 - 3 | 779 | 136 | ||
XM_546111.6 | XP_546111.3 | 2 - 3 | 775 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |