Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022414142.1 | XP_022269850.1 | 3 - 4 | 493 | 83 | |
XM_545302.5 | XP_545302.3 | 3 - 4 | 516 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |