Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080897.1 | NP_001074366.1 | 5 - 6 | 478 | 70 | |
XM_005636022.3 | XP_005636079.1 | 6 - 7 | 1121 | 70 | ||
XM_005636023.3 | XP_005636080.1 | 5 - 6 | 1004 | 70 | ||
GenBank mRNA | EF195231.1 | - | 5 - 6 | 478 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |