Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERTAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547396.6 | XP_547396.2 | 324 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |