Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTPCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014112738.2 | XP_013968213.2 | 320 | 109 | ||
XM_014112739.1 | XP_013968214.1 | 339 | 109 | |||
XM_022417412.1 | XP_022273120.1 | 531 | 109 | |||
XM_022417413.1 | XP_022273121.1 | 404 | 109 | |||
XM_843650.5 | XP_848743.3 | 321 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |