Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022418606.1 | XP_022274314.1 | 2 - 3 | 448 | 63 | |
XM_546637.5 | XP_546637.2 | 2 - 3 | 669 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |