Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASA4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620995.3 | XP_005621052.1 | 402 | 104 | ||
XM_014114180.2 | XP_013969655.1 | 402 | 104 | |||
XM_014114182.2 | XP_013969657.2 | 499 | 104 | |||
XM_022419592.1 | XP_022275300.1 | 499 | 104 | |||
XM_022419593.1 | XP_022275301.1 | 499 | 104 | |||
XM_022419594.1 | XP_022275302.1 | 499 | 104 | |||
XM_022419595.1 | XP_022275303.1 | 499 | 104 | |||
XM_022419597.1 | XP_022275305.1 | 402 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |