Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423299.1 | XP_022279007.1 | 3 - 4 | 647 | 63 | |
XM_022423300.1 | XP_022279008.1 | 3 - 4 | 670 | 63 | ||
XM_548256.6 | XP_548256.2 | 2 - 3 | 448 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |