Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287158.1 | NP_001274087.1 | 3 - 4 | 587 | 65 | |
GenBank mRNA | DN379133.1 | - | 3 - 4 | 587 | 65 | |
DN432131.1 | - | 2 - 3 | 225 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |