Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_843295.5 | XP_848388.1 | 6 - 7 | 822 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |