Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419553.1 | XP_022275261.1 | 478 | 74 | ||
XM_546921.6 | XP_546921.1 | 598 | 74 |