Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639517.3 | XP_005639574.1 | 3 - 4 | 482 | 118 | |
XM_005639518.2 | XP_005639575.1 | 3 - 4 | 431 | 118 | ||
XM_005639520.3 | XP_005639577.1 | 3 - 4 | 497 | 118 | ||
XM_005639521.2 | XP_005639578.1 | 3 - 4 | 841 | 118 | ||
XM_022414040.1 | XP_022269748.1 | 3 - 4 | 448 | 118 | ||
XM_022414041.1 | XP_022269749.1 | 3 - 4 | 507 | 118 | ||
XM_022414044.1 | XP_022269752.1 | 3 - 4 | 507 | 118 | ||
XM_022414045.1 | XP_022269753.1 | 3 - 4 | 391 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |