Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846899.4 | XP_851992.1 | 1 - 2 | 229 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |