Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_847118.4 | XP_852211.1 | 3 - 4 | 418 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |