Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111830.2 | XP_013967305.1 | 3 - 4 | 243 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |