Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111830.2 | XP_013967305.1 | 5 - 6 | 390 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |