Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413093.1 | XP_022268801.1 | 3 - 4 | 677 | 74 | |
XM_022413094.1 | XP_022268802.1 | 3 - 4 | 220 | 74 | ||
XM_022413095.1 | XP_022268803.1 | 3 - 4 | 253 | 74 | ||
XM_854663.6 | XP_859756.2 | 3 - 4 | 275 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |