Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413094.1 | XP_022268802.1 | 6 - 7 | 461 | 67 | |
XM_854663.6 | XP_859756.2 | 5 - 6 | 447 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |