Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618122.3 | XP_005618179.1 | 544 | 62 | ||
XM_014112225.2 | XP_013967700.1 | 416 | 62 | |||
XM_014112227.2 | XP_013967702.1 | 416 | 62 | |||
XM_022416728.1 | XP_022272436.1 | 487 | 62 | |||
XM_847057.5 | XP_852150.1 | 504 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris |