Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405409.1 | XP_022261117.1 | 8 - 9 | 1669 | 84 | |
XM_022405410.1 | XP_022261118.1 | 7 - 8 | 1534 | 84 | ||
XM_540610.6 | XP_540610.2 | 7 - 8 | 1498 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |