Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014121786.2 | XP_013977261.1 | 2 - 3 | 379 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |