Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845177.5 | XP_850270.1 | 3 - 4 | 731 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |