Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639577.4 | XP_003639625.1 | 1 - 2 | 298 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |