Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_848875.5 | XP_853968.1 | 1 - 2 | 196 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |