Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850300.5 | XP_855393.1 | 2 - 3 | 398 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |