Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014122801.2 | XP_013978276.1 | 4 - 5 | 342 | 95 | |
XM_022407942.1 | XP_022263650.1 | 4 - 5 | 342 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |