Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53I13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423398.1 | XP_022279106.1 | 1 - 2 | 189 | 90 | |
XM_022423399.1 | XP_022279107.1 | 1 - 2 | 185 | 90 | ||
XM_022423400.1 | XP_022279108.1 | 1 - 2 | 184 | 90 | ||
XM_848643.4 | XP_853736.1 | 1 - 2 | 186 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |