Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022411081.1 | XP_022266789.1 | 5 - 6 | 1276 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |