Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423258.1 | XP_022278966.1 | 2 - 3 | 916 | 76 | |
XM_847666.5 | XP_852759.2 | 2 - 3 | 917 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |