Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849018.5 | XP_854111.1 | 196 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |