Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022411026.1 | XP_022266734.1 | 3 - 4 | 507 | 82 | |
XM_849067.4 | XP_854160.3 | 3 - 4 | 505 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |